Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017802.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017802.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017802.hg.1 |
TC16001278.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017803.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017803.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017803.hg.1 |
TC16001278.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000516313.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000564793.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000569341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000569341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000569341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000570279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000612761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000612761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
ENST00000612761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017804.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017804.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16017804.hg.1 |
TC16001278.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194439.hg.1 |
TC1600011565.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600194441.hg.1 |
TC1600011565.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000283882.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000516313.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000562602.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564286.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000564793.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000565646.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000566901.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000569341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000569341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000569341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000570279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000612761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000612761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
ENST00000612761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011565.hg.1 |
NM_006324.2 |
Refseq |