Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007325.hg.1 |
ENST00000469935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007325.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14007325.hg.1 |
TC14000588.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000478994.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000478994.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
ENST00000478994.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400157335.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400157335.hg.1 |
ENST00000478994.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400157335.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400157335.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400157335.hg.1 |
ENST00000478994.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400157335.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400157335.hg.1 |
TC1400008070.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000469935.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000478994.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000478994.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
ENST00000478994.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008070.hg.1 |
NR_015340.1 |
Refseq |