Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006637.hg.1 |
ENST00000200676.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006637.hg.1 |
ENST00000379780.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006637.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16006637.hg.1 |
TC16000481.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007985.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
ENST00000200676.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
ENST00000379780.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
ENST00000566128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
ENST00000569082.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
NM_000078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
NM_001286085.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600158379.hg.1 |
TC1600007985.hg.1 |
Clariom D TC |