Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003664.hg.1 |
TC01000231.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003664.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003664.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003666.hg.1 |
TC01000231.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003666.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003666.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003665.hg.1 |
TC01000231.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003665.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003665.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149557.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149557.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149557.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149557.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149557.hg.1 |
TC0100007103.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100149559.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149559.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149559.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149559.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149559.hg.1 |
TC0100007103.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007103.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149556.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149556.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149556.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149556.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149556.hg.1 |
TC0100007103.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100149558.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149558.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149558.hg.1 |
ENST00000375460.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100149558.hg.1 |
NM_016233.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100149558.hg.1 |
TC0100007103.hg.1 |
Clariom D TC |