Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034865.hg.1 |
TC01002287.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034865.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034865.hg.1 |
ENST00000375486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034865.hg.1 |
ENST00000444885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034865.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034865.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034866.hg.1 |
TC01002287.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034866.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034866.hg.1 |
ENST00000375486.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034866.hg.1 |
ENST00000444885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034866.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01034866.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
ENST00000479534.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013076.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196654.hg.1 |
TC0100013076.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100196652.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196652.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196652.hg.1 |
TC0100013076.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
ENST00000375481.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
ENST00000375486.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
ENST00000466151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
NM_007365.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100196653.hg.1 |
TC0100013076.hg.1 |
Clariom D TC |