Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024186.hg.1 |
TC06001963.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024186.hg.1 |
ENST00000254759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024186.hg.1 |
ENST00000369242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024186.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024186.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024185.hg.1 |
TC06001963.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024185.hg.1 |
ENST00000254759.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024185.hg.1 |
ENST00000369240.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024185.hg.1 |
ENST00000369242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024185.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024185.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187166.hg.1 |
TC0600012654.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012654.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
ENST00000254759.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
ENST00000369240.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
ENST00000369242.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
ENST00000479163.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
NM_017421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600187165.hg.1 |
TC0600012654.hg.1 |
Clariom D TC |