Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026616.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026616.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026616.hg.1 |
TC11002228.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000531200.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012123.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193141.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193141.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193141.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193141.hg.1 |
ENST00000260227.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193141.hg.1 |
ENST00000533366.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193141.hg.1 |
NM_002423.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193141.hg.1 |
TC1100012123.hg.1 |
Clariom D TC |