Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031208.3 | NP_112485.1 | 1 | 1315 | 199 | |
GenBank mRNA | AF293367.1 | - | 1 | 344 | 199 | |
AK094199.1 | - | 1 | 1314 | 199 | ||
AL136720.1 | - | 1 | 1316 | 199 | ||
BC020615.1 | - | 1 | 157 | 199 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |