Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036.4 | NP_001027.3 | 84 - 85 | 11334 | 100 | |
NM_001243996.2 | NP_001230925.1 | 83 - 84 | 11319 | 100 | ||
XM_011521880.2 | XP_011520182.1 | 82 - 83 | 11489 | 100 | ||
XM_017022468.1 | XP_016877957.1 | 83 - 84 | 11510 | 100 | ||
XM_017022469.1 | XP_016877958.1 | 83 - 84 | 11507 | 100 | ||
XM_017022470.1 | XP_016877959.1 | 83 - 84 | 13844 | 100 | ||
XM_017022471.1 | XP_016877960.1 | 83 - 84 | 11507 | 100 | ||
XM_017022472.1 | XP_016877961.1 | 83 - 84 | 11504 | 100 | ||
XM_017022473.1 | XP_016877962.1 | 82 - 83 | 11492 | 100 | ||
XM_017022474.1 | XP_016877963.1 | 83 - 84 | 11474 | 100 | ||
XM_017022475.1 | XP_016877964.1 | 83 - 84 | 11510 | 100 | ||
XM_017022476.1 | XP_016877965.1 | 82 - 83 | 11402 | 100 | ||
XM_017022477.1 | XP_016877966.1 | 83 - 84 | 11511 | 100 | ||
GenBank mRNA | AB001025.1 | - | 83 - 84 | 11324 | 100 | |
AJ001515.1 | - | 84 - 85 | 11320 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Detect Genomic DNA Homo sapiens Probe spans exons |