Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017169.hg.1 |
ENST00000299441.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017169.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017169.hg.1 |
TC11001363.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009982.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175457.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100175457.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175457.hg.1 |
ENST00000299441.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100175457.hg.1 |
NM_003737.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100175457.hg.1 |
TC1100009982.hg.1 |
Clariom D TC |