Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001207025.2 | NP_001193954.1 | 14 | 6462 | 67 | |
NM_001247994.1 | NP_001234923.1 | 14 | 6278 | 67 | ||
NM_002698.4 | NP_002689.1 | 14 | 6414 | 67 | ||
XM_005259010.3 | XP_005259067.1 | 15 | 6505 | 67 | ||
XM_011527040.2 | XP_011525342.1 | 15 | 6619 | 67 | ||
XM_011527041.2 | XP_011525343.1 | 14 | 6545 | 67 | ||
XM_011527042.2 | XP_011525344.1 | 14 | 6497 | 67 | ||
XM_011527043.2 | XP_011525345.2 | 15 | 6706 | 67 | ||
XM_017026884.1 | XP_016882373.1 | 16 | 6917 | 67 | ||
XM_017026885.1 | XP_016882374.1 | 16 | 6920 | 67 | ||
XM_017026886.1 | XP_016882375.1 | 17 | 8419 | 67 | ||
XM_017026887.1 | XP_016882376.1 | 17 | 6889 | 67 | ||
XM_017026888.1 | XP_016882377.1 | 17 | 7369 | 67 | ||
XM_017026889.1 | XP_016882378.1 | 16 | 6911 | 67 | ||
XM_017026890.1 | XP_016882379.1 | 16 | 6992 | 67 | ||
XM_017026891.1 | XP_016882380.1 | 16 | 6869 | 67 | ||
XM_017026892.1 | XP_016882381.1 | 16 | 6851 | 67 | ||
XM_017026893.1 | XP_016882382.1 | 15 | 6571 | 67 | ||
XM_017026894.1 | XP_016882383.1 | 15 | 6553 | 67 | ||
XM_017026895.1 | XP_016882384.1 | 15 | 6551 | 67 | ||
XM_017026896.1 | XP_016882385.1 | 15 | 6843 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |