Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK126751.1 | - | 1 | 3324 | 139 | |
BC013743.1 | - | 1 | 2741 | 139 | ||
BX647663.1 | - | 1 | 3828 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |