Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194839.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194839.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194839.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200194839.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200194839.hg.1 |
TC1200012159.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000257552.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546622.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
ENST00000546985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012159.hg.1 |
NM_002442.3 |
Refseq |