Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011529820.2 | XP_011528122.1 | 2 - 3 | 206 | 63 | |
XM_017028530.1 | XP_016884019.1 | 2 - 3 | 206 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |