Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000315994.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000355946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000369774.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
ENST00000420222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011726.hg.1 |
NM_014631.2 |
Refseq |