Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004815.hg.1 |
ENST00000282028.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004815.hg.1 |
ENST00000326695.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004815.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004815.hg.1 |
TC18000470.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000594852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
ENST00000594852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
TC1800008549.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800158240.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000326695.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000589109.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000590910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000594852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
ENST00000594852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_001272077.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008549.hg.1 |
NM_002930.3 |
Refseq |