Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
TC2200006654.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000356671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000357502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000400451.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000403682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000405993.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000437275.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000476678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000493734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000611540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NM_001256523.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NM_001256524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NM_001256525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NM_003426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NR_046282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000356671.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000357502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000400451.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000403682.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000405993.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000437275.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000476678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000493734.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
ENST00000611540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NM_001256523.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NM_001256524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NM_001256525.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NM_003426.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200145705.hg.1 |
NR_046282.1 |
Refseq |