Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135022.1 | NP_001128494.1 | 3 - 4 | 250 | 219 | |
XM_005264596.3 | XP_005264653.1 | 3 - 4 | 250 | 219 | ||
XM_005264598.3 | XP_005264655.1 | 2 - 3 | 155 | 124 | ||
XM_017005078.1 | XP_016860567.1 | 2 - 3 | 106 | 124 | ||
GenBank mRNA | BC010991.2 | - | 3 - 4 | 182 | 3936410 | 219 |
DA294897.1 | - | 2 - 3 | 159 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |