Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP7D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286365.1 | NP_001273294.1 | 13 - 14 | 2569 | 72 | |
NM_001286366.1 | NP_001273295.1 | 15 - 16 | 2584 | 72 | ||
XM_006710759.1 | XP_006710822.1 | 14 - 15 | 2495 | 72 | ||
XM_006710761.1 | XP_006710824.1 | 14 - 15 | 2288 | 72 | ||
GenBank mRNA | AB033013.2 | - | 13 - 14 | 2569 | 72 | |
AF218002.1 | - | 14 - 15 | 1907 | 73 | ||
AK001212.1 | - | 6 - 7 | 763 | 72 | ||
AK095939.1 | - | 7 - 8 | 927 | 72 | ||
AK096341.1 | - | 15 - 16 | 2351 | 72 | ||
AK309966.1 | - | 12 - 13 | 1575 | 72 | ||
AL136547.1 | - | 6 - 7 | 594 | 72 | ||
BC003083.2 | - | 6 - 7 | 830 | 72 | ||
BC027334.1 | - | 4 - 5 | 355 | 72 | ||
BC057293.1 | - | 13 - 14 | 2371 | 72 | ||
BC067256.1 | - | 11 - 12 | 1742 | 72 | ||
BC082266.1 | - | 8 - 9 | 1105 | 72 | ||
BC128042.1 | - | 4 - 5 | 201 | 72 | ||
CR457254.1 | - | 5 - 6 | 460 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |