Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNPY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318845.1 | NP_001305774.1 | 1 - 2 | 587 | 86 | |
NR_134886.1 | 1 - 2 | 587 | 86 | |||
NR_134888.1 | 1 - 2 | 587 | 86 | |||
GenBank mRNA | BC008133.1 | - | 1 - 2 | 223 | 86 | |
BF528267.1 | - | 1 - 2 | 179 | 86 | ||
HY101691.1 | - | 1 - 2 | 235 | 86 |
Gene Symbol: CNPY3-GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134890.1 | 1 - 2 | 587 | 86 | ||
GenBank mRNA | FY213816.1 | - | 1 - 2 | 119 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |