Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007184.hg.1 |
TC07000508.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007184.hg.1 |
ENST00000351004.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007184.hg.1 |
ENST00000457358.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007184.hg.1 |
ENST00000490206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007184.hg.1 |
NM_001256414.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07007184.hg.1 |
NM_002069.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000351004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000418742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000447650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000457358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000490206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
NM_001256414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
NM_002069.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000351004.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000418742.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000447650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000457358.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
ENST00000490206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
NM_001256414.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
NM_002069.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700156867.hg.1 |
TC0700008181.hg.1 |
Clariom D TC |