Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
ENST00000329667.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
ENST00000360525.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
ENST00000399098.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
ENST00000399102.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
ENST00000399103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
ENST00000464265.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005143.hg.1 |
TC21000439.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
TC2100008095.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154641.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000329667.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000360525.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399098.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399102.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000399103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000430792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000464265.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
ENST00000479152.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NM_153682.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008095.hg.1 |
NR_131189.1 |
Refseq |