Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17027038.hg.1 |
ENST00000312648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17027038.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17027038.hg.1 |
TC17001978.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012109.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700199524.hg.1 |
TC1700012109.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000312648.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000578509.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000581434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000581744.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000582480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000583376.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
ENST00000584284.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
NM_006137.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700199525.hg.1 |
TC1700012109.hg.1 |
Clariom D TC |