Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011543537.2 | XP_011541839.1 | 2 - 3 | 203 | 85 | |
XM_017009698.1 | XP_016865187.1 | 1 - 2 | 120 | 85 | ||
XM_017009699.1 | XP_016865188.1 | 2 - 3 | 203 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |