Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005203.hg.1 |
ENST00000304718.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005203.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17005203.hg.1 |
TC17000438.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000612563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000612563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000612563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000612563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000615952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000615952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000615952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000620927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000620927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000620927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
ENST00000620927.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000611219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000612563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000612563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000612563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000612563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000615952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000615952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000615952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000616019.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000620927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000620927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000620927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
ENST00000620927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007638.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
ENST00000613102.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
ENST00000617860.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
ENST00000618837.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
ENST00000619573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
NM_024835.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155078.hg.1 |
TC1700007638.hg.1 |
Clariom D TC |