Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011027.hg.1 |
TC0X000805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011027.hg.1 |
ENST00000217890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011027.hg.1 |
ENST00000381154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011027.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011028.hg.1 |
TC0X000805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011028.hg.1 |
ENST00000217890.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011028.hg.1 |
ENST00000381154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011028.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000481340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000481340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011023.hg.1 |
TC0X000805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011023.hg.1 |
ENST00000381154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011023.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011024.hg.1 |
TC0X000805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011024.hg.1 |
ENST00000381154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011024.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011022.hg.1 |
TC0X000805.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011022.hg.1 |
ENST00000381154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X011022.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000481340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000481340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000494870.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000495294.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
ENST00000559324.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011338.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178356.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178356.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178356.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178356.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178356.hg.1 |
TC0X00011338.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00178357.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178357.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178357.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178357.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178357.hg.1 |
TC0X00011338.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00178355.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178355.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178355.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178355.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178355.hg.1 |
ENST00000458014.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178355.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178355.hg.1 |
TC0X00011338.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00178360.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178360.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178360.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178360.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178360.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178360.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178360.hg.1 |
TC0X00011338.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
ENST00000481340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
ENST00000217890.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
ENST00000381154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
ENST00000481340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
NM_001669.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00178361.hg.1 |
TC0X00011338.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006626.hg.1 |
ENST00000443820.1 |
Ensembl |