Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004646.3 | NP_004637.1 | 23 - 24 | 3322 | 145 | |
GenBank mRNA | AF035835.1 | - | 23 - 24 | 3166 | 145 | |
AF190637.1 | - | 23 - 24 | 3166 | 145 | ||
EU642886.1 | - | 23 - 24 | 3166 | 145 | ||
EU642887.1 | - | 23 - 24 | 3165 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |