Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004034.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004034.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004034.hg.1 |
TC16000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004035.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004035.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16004035.hg.1 |
TC16000285.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
ENST00000475436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
ENST00000475436.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152462.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152462.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152462.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152462.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152462.hg.1 |
TC1600007285.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600152461.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152461.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152461.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152461.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152461.hg.1 |
TC1600007285.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
ENST00000331351.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
ENST00000475436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
ENST00000475436.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007285.hg.1 |
NM_006040.2 |
Refseq |