Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030097.hg.1 |
TC02001889.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030097.hg.1 |
ENST00000316752.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030097.hg.1 |
ENST00000407787.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030097.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000398658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000398658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000398658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000398658.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000421319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000421319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000421319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000421319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000421319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200192755.hg.1 |
TC0200012727.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000316752.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000398658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000398658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000398658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000398658.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000407787.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000421319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000421319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000421319.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000430495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
ENST00000602599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012727.hg.1 |
NM_144709.3 |
Refseq |