Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006453.2 | NP_006444.2 | 20 - 21 | 2239 | 73 | |
GenBank mRNA | AB209173.1 | - | 20 - 21 | 2211 | 73 | |
AK095076.1 | - | 12 - 13 | 2415 | 73 | ||
BC007620.1 | - | 12 - 13 | 1423 | 73 | ||
BC010231.1 | - | 20 - 21 | 2239 | 3455740 | 73 | |
BC014007.2 | - | 20 - 21 | 2211 | 4040312 | 73 | |
BC035409.1 | - | 20 - 21 | 2195 | 5165833 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |