Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000547.hg.1 |
TC07000036.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000547.hg.1 |
ENST00000258796.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000547.hg.1 |
ENST00000403167.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000547.hg.1 |
ENST00000407643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07000547.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000400376.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000400376.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000400376.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000477439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000477439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000477439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000477439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700145127.hg.1 |
TC0700006567.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000258796.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000400376.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000400376.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000400376.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000403167.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000407643.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000429448.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000477439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000477439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000477439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000477439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
ENST00000498454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006567.hg.1 |
NM_025250.2 |
Refseq |