Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000528.3 | NP_000519.2 | 20 - 21 | 2512 | 70 | |
NM_001173498.1 | NP_001166969.1 | 20 - 21 | 2509 | 70 | ||
XM_005259913.1 | XP_005259970.1 | 20 - 21 | 2493 | 70 | ||
XM_017026818.1 | XP_016882307.1 | 17 - 18 | 2091 | 70 | ||
GenBank mRNA | AB209921.1 | - | 20 - 21 | 2424 | 70 | |
AK291572.1 | - | 20 - 21 | 2477 | 70 | ||
AK303418.1 | - | 21 - 22 | 2451 | 70 | ||
BC000736.2 | - | 20 - 21 | 2458 | 3504417 | 70 | |
U05572.1 | - | 20 - 21 | 2466 | 70 | ||
U60266.1 | - | 20 - 21 | 2450 | 70 | ||
U68382.1 | - | 18 - 19 | 3025 | 70 | ||
U68567.1 | - | 20 - 21 | 2424 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |