Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023187.hg.1 |
ENST00000225504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023187.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023187.hg.1 |
TC17001731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023188.hg.1 |
ENST00000225504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023188.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023188.hg.1 |
TC17001731.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700206893.hg.1 |
TC1700012438.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000225504.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000577396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000579289.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581166.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
ENST00000581540.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NM_003168.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012438.hg.1 |
NR_073470.1 |
Refseq |