Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193329.1 | NP_001180258.1 | 6 - 7 | 1788 | 75 | |
XM_006717306.1 | XP_006717369.1 | 7 - 8 | 1936 | 75 | ||
XM_011519119.1 | XP_011517421.1 | 7 - 8 | 1936 | 75 | ||
XM_011519120.2 | XP_011517422.1 | 7 - 8 | 1936 | 75 | ||
XM_011519121.2 | XP_011517423.1 | 7 - 8 | 1936 | 75 | ||
XM_011519122.2 | XP_011517424.1 | 7 - 8 | 1936 | 75 | ||
XM_011519123.2 | XP_011517425.1 | 7 - 8 | 1936 | 75 | ||
XM_011519127.1 | XP_011517429.1 | 7 - 8 | 1936 | 75 | ||
XM_011519132.1 | XP_011517434.1 | 4 - 5 | 662 | 75 | ||
XM_017015229.1 | XP_016870718.1 | 7 - 8 | 1979 | 75 | ||
XM_017015230.1 | XP_016870719.1 | 7 - 8 | 1936 | 75 | ||
XM_017015232.1 | XP_016870721.1 | 7 - 8 | 1936 | 75 | ||
XM_017015233.1 | XP_016870722.1 | 7 - 8 | 1966 | 75 | ||
XM_017015234.1 | XP_016870723.1 | 6 - 7 | 1769 | 75 | ||
XM_017015235.1 | XP_016870724.1 | 6 - 7 | 1769 | 75 | ||
GenBank mRNA | AJ560639.1 | - | 6 - 7 | 1653 | 75 | |
BC020194.1 | - | 6 - 7 | 999 | 5020674 | 75 | |
BX648970.1 | - | 5 - 6 | 874 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |