Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000098.hg.1 |
ENST00000400572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000098.hg.1 |
ENST00000482980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000098.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000098.hg.1 |
TC21000026.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000429114.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000471902.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006586.hg.1 |
NR_003088.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
NR_003087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
TC2100006586.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
ENST00000463099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
ENST00000467409.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
ENST00000481582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
ENST00000482980.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100144771.hg.1 |
NR_003087.1 |
Refseq |