Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000330622.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000427718.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000547806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000547806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000547806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000548840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000548840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000548840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000548840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000548840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000548840.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000549689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000549689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000549689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000549689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000549689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000549689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000549689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000550180.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000551458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000551458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000551458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000551458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000551458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000551458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000551458.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
ENST00000622495.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_015002.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012083.hg.1 |
NM_033624.2 |
Refseq |