Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020286.hg.1 |
TC09002920.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020286.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020286.hg.1 |
ENST00000373156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020286.hg.1 |
ENST00000373176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020286.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000413016.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550143.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012277.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
ENST00000223836.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
ENST00000373156.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
ENST00000373176.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
ENST00000550992.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
NM_000476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186714.hg.1 |
TC0900012277.hg.1 |
Clariom D TC |