Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019235.hg.1 |
ENST00000358334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019235.hg.1 |
ENST00000359263.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019235.hg.1 |
ENST00000371501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019235.hg.1 |
ENST00000371502.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019235.hg.1 |
ENST00000463541.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019235.hg.1 |
NM_013451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019235.hg.1 |
NM_133337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019235.hg.1 |
TC10001535.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019236.hg.1 |
ENST00000358334.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019236.hg.1 |
ENST00000359263.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019236.hg.1 |
ENST00000371501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019236.hg.1 |
ENST00000371502.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019236.hg.1 |
NM_013451.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019236.hg.1 |
NM_133337.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019236.hg.1 |
TC10001535.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
ENST00000358334.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
ENST00000359263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
ENST00000463743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
NM_013451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
NM_133337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
ENST00000358334.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
ENST00000359263.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
ENST00000463743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
NM_013451.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
NM_133337.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177644.hg.1 |
TC1000011445.hg.1 |
Clariom D TC |