Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014720.3 | NP_055535.2 | 13 - 14 | 3437 | 80 | |
XM_011540401.2 | XP_011538703.1 | 12 - 13 | 2093 | 80 | ||
GenBank mRNA | AF273048.1 | - | 13 - 14 | 2910 | 80 | |
EF139853.1 | - | 3 - 4 | 234 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Homo sapiens Probe spans exons |