Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000473775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
ENST00000473775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006768.hg.1 |
TC13001719.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006768.hg.1 |
ENST00000379561.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006768.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013117.hg.1 |
ENST00000504678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006769.hg.1 |
TC13001719.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006769.hg.1 |
ENST00000379561.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006769.hg.1 |
ENST00000473775.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006769.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006772.hg.1 |
TC13001719.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006772.hg.1 |
ENST00000379561.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006772.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006773.hg.1 |
TC13001719.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006773.hg.1 |
ENST00000379561.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006773.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158672.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158672.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158672.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158672.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158672.hg.1 |
TC1300008688.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000473775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
ENST00000473775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008688.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158673.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158673.hg.1 |
ENST00000473775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158673.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158673.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158673.hg.1 |
ENST00000473775.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158673.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158673.hg.1 |
TC1300008688.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300158675.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158675.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158675.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158675.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158675.hg.1 |
TC1300008688.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300158676.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158676.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158676.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158676.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158676.hg.1 |
TC1300008688.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300158677.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158677.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158677.hg.1 |
ENST00000379561.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158677.hg.1 |
NM_002015.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158677.hg.1 |
TC1300008688.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500013117.hg.1 |
ENST00000504678.1 |
Ensembl |