Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000522105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000522105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000522105.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007394.hg.1 |
TC08000542.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007394.hg.1 |
ENST00000518476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007394.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800156018.hg.1 |
TC0800008154.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000517748.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000518476.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000521593.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000522105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000522105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000522105.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000522427.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000523299.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
ENST00000620844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008154.hg.1 |
NM_173538.2 |
Refseq |