Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000379727.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395048.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000395049.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000562201.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000564596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000566503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000567032.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000569630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000612821.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
ENST00000617691.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010042.hg.1 |
NM_000499.4 |
Refseq |