Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029957.hg.1 |
TC07001927.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029957.hg.1 |
ENST00000288602.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029957.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029955.hg.1 |
TC07001927.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029955.hg.1 |
ENST00000288602.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029955.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700202225.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700202225.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700202225.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700202225.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700202225.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700202225.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700202225.hg.1 |
TC0700013629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700202229.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700202229.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700202229.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700202229.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700202229.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700202229.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700202229.hg.1 |
TC0700013629.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000288602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000469930.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000479537.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
ENST00000497784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013629.hg.1 |
NM_004333.5 |
Refseq |