Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000608879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046866.hg.1 |
TC01003010.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046866.hg.1 |
ENST00000369545.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046866.hg.1 |
ENST00000369546.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046866.hg.1 |
ENST00000369547.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046866.hg.1 |
ENST00000393296.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046866.hg.1 |
ENST00000483203.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046866.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608203.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000608879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000609577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015342.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000369547.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000607941.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000609117.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000610096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000610121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
ENST00000610222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
NM_001270805.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
NM_205848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214556.hg.1 |
TC0100015342.hg.1 |
Clariom D TC |