Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000995.hg.1 |
ENST00000378827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000995.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000995.hg.1 |
TC20000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000997.hg.1 |
ENST00000378827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000997.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000997.hg.1 |
TC20000067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006642.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145555.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145555.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145555.hg.1 |
TC2000006642.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145555.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145555.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145552.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145552.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000145552.hg.1 |
TC2000006642.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000145552.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000145552.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
ENST00000378827.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
NM_001200.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006642.hg.1 |
NM_001200.3 |
Refseq |