Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015082.hg.1 |
ENST00000299164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015082.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015082.hg.1 |
TC11001197.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015083.hg.1 |
ENST00000299164.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015083.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015083.hg.1 |
TC11001197.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171689.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171689.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171689.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171689.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171689.hg.1 |
TC1100009533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100171688.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171688.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171688.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171688.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171688.hg.1 |
TC1100009533.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
ENST00000299164.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009533.hg.1 |
NM_139055.3 |
Refseq |