Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014614.2 | NP_055429.2 | 25 - 26 | 2906 | 152 | |
XM_006711969.1 | XP_006712032.1 | 24 - 25 | 2483 | 152 | ||
XM_011532705.2 | XP_011531007.1 | 26 - 27 | 2930 | 152 | ||
XM_011532706.2 | XP_011531008.1 | 26 - 27 | 2930 | 152 | ||
XM_011532707.2 | XP_011531009.1 | 25 - 26 | 2864 | 152 | ||
XM_011532709.2 | XP_011531011.1 | 26 - 27 | 2930 | 152 | ||
GenBank mRNA | AB621805.1 | - | 24 - 25 | 3023 | 152 | |
AY894754.1 | - | 24 - 25 | 2755 | 152 | ||
AY894755.1 | - | 6 - 7 | 594 | 152 | ||
BC112169.1 | - | 24 - 25 | 2542 | 8327637 | 152 | |
BC113668.1 | - | 24 - 25 | 2542 | 8322720 | 152 | |
BC143739.1 | - | 24 - 25 | 2542 | 152 | ||
D38521.1 | - | 25 - 26 | 2716 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |