Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020106.hg.1 |
ENST00000355468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020106.hg.1 |
ENST00000393928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020106.hg.1 |
ENST00000545545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020106.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020106.hg.1 |
TC17001517.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020105.hg.1 |
ENST00000355468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020105.hg.1 |
ENST00000393928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020105.hg.1 |
ENST00000545545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020105.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020105.hg.1 |
TC17001517.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000465097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000465097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000465097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000465097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000590496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000590496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000590496.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184678.hg.1 |
TC1700010687.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000355468.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000393928.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000465097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000465097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000465097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000465097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000467164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000484247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000587455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000590496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000590496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000590496.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
ENST00000592026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010687.hg.1 |
NM_006455.2 |
Refseq |