Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198934.1 | NP_001185863.1 | 20 - 21 | 4418 | 72 | |
NM_033450.2 | NP_258261.2 | 18 - 19 | 4478 | 72 | ||
XM_011514974.2 | XP_011513276.2 | 19 - 20 | 4403 | 72 | ||
XM_011514983.2 | XP_011513285.1 | 20 - 21 | 3280 | 72 | ||
XM_011514985.2 | XP_011513287.1 | 14 - 15 | 2619 | 72 | ||
XM_011514986.2 | XP_011513288.1 | 17 - 18 | 2575 | 72 | ||
XM_017011445.1 | XP_016866934.1 | 19 - 20 | 4449 | 72 | ||
XM_017011446.1 | XP_016866935.1 | 20 - 21 | 4479 | 72 | ||
XM_017011447.1 | XP_016866936.1 | 20 - 21 | 4421 | 72 | ||
XM_017011450.1 | XP_016866939.1 | 19 - 20 | 4435 | 72 | ||
XM_017011452.1 | XP_016866941.1 | 21 - 22 | 4597 | 72 | ||
XM_017011454.1 | XP_016866943.1 | 19 - 20 | 3087 | 72 | ||
XM_017011455.1 | XP_016866944.1 | 19 - 20 | 3061 | 72 | ||
XM_017011456.1 | XP_016866945.1 | 15 - 16 | 2398 | 72 | ||
GenBank mRNA | AK000002.1 | - | 20 - 21 | 4418 | 72 | |
AK024446.1 | - | 14 - 15 | 3931 | 72 | ||
AL122095.1 | - | 3 - 4 | 375 | 72 | ||
AL133613.1 | - | 11 - 12 | 1829 | 72 | ||
AY032599.1 | - | 18 - 19 | 4474 | 72 | ||
BC024103.1 | - | 5 - 6 | 679 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |