Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023642.hg.1 |
TC07001566.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023642.hg.1 |
ENST00000307792.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023642.hg.1 |
ENST00000427262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023642.hg.1 |
ENST00000541514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023642.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07023642.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700183227.hg.1 |
TC0700011658.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000307792.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000427262.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000442159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
ENST00000453333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_001178129.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700011658.hg.1 |
NM_012431.2 |
Refseq |